Learn More
Waters Corp ACQUITY UPLC Method Development Kit, 130Å, 1.7 µm, 2.1 mm X 50 mm, 4/pk
Supplier:  Waters Corp 176001042
Encompass_Preferred
ACQUITY UPLC columns are the most technologically advanced LC columns ever created and are the only columns designed, tested and certified to withstand the pressures of UPLC Technology. Based upon the highly efficient 1.7 µm Bridged Ethyl Hybrid (BEH) particles and 1.8 µm High Strength Silica (HSS) particles, these columns are the first and only columns that allow separation scientists to achieve maximum speed, sensitivity and resolution without compromise. The particles and chemistries of the ACQUITY UPLC column family were carefully chosen to produce the ideal combination of ultra-performance efficiencies, wide pH range and complementary selectivities. ACQUITY UPLC BEH columns and ACQUITY UPLC HSS columns are the only columns recommended for use with ACQUITY UPLC Systems.
Catalog No. 50-780-017
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.