Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine Zone Refined (number of passes:45) 99.0+%, TCI America™

Click to view available options
Quantity:
ZONE
260-94-6
C13H9N
179.222
MFCD00005025
DZBUGLKDJFMEHC-UHFFFAOYSA-N
9-azaanthracene,2,3-benzoquinoline,acrydine,10-azaanthracene,akridin,dibenzo b,e pyridine,2,3,5,6-dibenzopyridine,akridin czech,benzo b quinoline
9215
CHEBI:36420
acridine
C1=CC=C2C(=C1)C=C3C=CC=CC3=N2
260-94-6
C13H9N
179.222
MFCD00005025
DZBUGLKDJFMEHC-UHFFFAOYSA-N
9-azaanthracene,2,3-benzoquinoline,acrydine,10-azaanthracene,akridin,dibenzo b,e pyridine,2,3,5,6-dibenzopyridine,akridin czech,benzo b quinoline
9215
CHEBI:36420
acridine
C1=CC=C2C(=C1)C=C3C=CC=CC3=N2
Melting Point 110°C
Quantity ZONE
Color Undesignated
UN Number 2713
Formula Weight 179.22
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (HPLC,T)
Chemical Name or Material Acridine Zone Refined (number of passes:45)

RTECSNumber : AR7175000

TSCA : Yes