Learn More
Medchemexpress LLC Acridone | 578-95-0 | 195.22 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00777150G
Encompass_Preferred
Acridone is an organic compound based on the acridine skeleton, recognized for its antibacterial, antimalarial, antiviral, and antineoplastic activities. This light yellow to green yellow solid is intended for research use only and inhibits the proliferation of MDA-MB-231 cancer cells, while also blocking the mRNA and protein expression of ATP binding cassette subfamily G member 2 (ABCG2).
  • Organic compound based on acridine skeleton
  • Possesses antibacterial activity
  • Exhibits antimalarial activity
  • Demonstrates antiviral activity
  • Shows antineoplastic activities
  • Inhibits proliferation of cancer cell MDA-MB-231
  • Blocks mRNA and protein expression of ABCG2
  • Appears as a light yellow to green yellow solid
Catalog No. 50-004-46532