Learn More
Cytiva Acro 37 TF Vent Device with PTFE Membrane - 0.2 µm, 37 mm (24/pkg)
Supplier:  Cytiva 4464
Encompass_Preferred
Disposable, multipurpose filters for small volume venting, solvent, and gas filtration.-Hydrophobic membrane acts as a barrier to aqueous solutions and aerosols-Protect sensitive equipment or the laboratory environment-Help vent small volume culture vessels in in-line filtration of air and other gasses-Have Broad chemical compatibility for solvent filtrationComes with PTFE membrane and polypropylene housing. Membrane has 0.2µm pore size and 37mm diameter with effective filtration area of 7.5cm2. Hose barb connectors are 6.4–9.5mm (1/4–3/8"). Maximum operating temperature: 100°C (212°F). Maximum operating pressure: 4.1bar (60psi), bidirectional. Nonsterile. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0417062
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.