Learn More
Andwin ScientificSupplier Diversity Partner ACRO 50 VENT FLT EMFLON 3/PK DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner A50V002P2
The 0.2 �m Acro� vent filter can be used for several laboratory procedures, including venting of bioreactors, fermentation tanks, and carboys. It is autoclavable at the temperature of 121�C (250�F) for 20 minutes. This product is nonsterile and is disposable. It is sold in a pack of 3.
Catalog No. NC0000081