Learn More
Andwin ScientificSupplier Diversity Partner ACRODISC .2 UM MINISPI 1000/PK DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 4567
The Acrodisc� PSF syringe filter is a multi-layered prefiltration device with an automation-certified filter housing. It is an efficient laboratory accessory for automated sampling or sample clarification. With a glass fiber prefilter version, this product allows filtration of thick, particulate-laden samples. It is sold in a pack of 1000 filters with a pore size of 0.2 um.
Catalog No. NC0215063