Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrolein 2,4-Dinitrophenylhydrazone 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
888-54-0
C9H8N4O4
236.187
MFCD00191599
JJPZHGIYUVFTGG-BJMVGYQFSA-N
acrolein 2,4-dinitrophenylhydrazone,acrolein-2,4-dinitrophenylhydrazone,2-propenal, 2,4-dinitrophenyl hydrazone,acrolein-2,4-dinitrophenylhydrazone dnph,2,4-dinitro-n-e-prop-2-enylideneamino aniline,1e-1-azabuta-1,3-dienyl 2,4-dinitrophenyl amine,1-allylidene-2-2,4-dinitrophenyl hydrazine,2,4-dinitro-n-prop-2-enylideneamino aniline,2-propenal, 2-2,4-dinitrophenyl hydrazone,acrolein, 2,4-dinitrophenyl hydrazone 8ci
9568683
2,4-dinitro-N-[(E)-prop-2-enylideneamino]aniline
C=CC=NNC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]
888-54-0
C9H8N4O4
236.187
MFCD00191599
JJPZHGIYUVFTGG-BJMVGYQFSA-N
acrolein 2,4-dinitrophenylhydrazone,acrolein-2,4-dinitrophenylhydrazone,2-propenal, 2,4-dinitrophenyl hydrazone,acrolein-2,4-dinitrophenylhydrazone dnph,2,4-dinitro-n-e-prop-2-enylideneamino aniline,1e-1-azabuta-1,3-dienyl 2,4-dinitrophenyl amine,1-allylidene-2-2,4-dinitrophenyl hydrazine,2,4-dinitro-n-prop-2-enylideneamino aniline,2-propenal, 2-2,4-dinitrophenyl hydrazone,acrolein, 2,4-dinitrophenyl hydrazone 8ci
9568683
2,4-dinitro-N-[(E)-prop-2-enylideneamino]aniline
C=CC=NNC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]
Color Undesignated
Quantity 1g
Melting Point 165°C
UN Number 1325
Formula Weight 236.19
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Acrolein 2,4-Dinitrophenylhydrazone