Acrolein, 96%, stab. with hydroquinone, Alfa Aesar

Manufacturer: Alfa Aesar 36520AE

Quantity 100mL

 View more versions of this product

Catalog No. AA36520AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107-02-8
    Physical Form Liquid
    Assay Percent Range 96%
    Molecular Formula H2C=CHCHO
    Formula Weight 56.06
    Chemical Name or Material Acrolein

    Specifications

    Formula Weight 56.06
    Quantity 100mL
    Physical Form Liquid
    Density 0.84
    Assay Percent Range 96%
    Molecular Formula H2C=CHCHO
    CAS 107-02-8
    Chemical Name or Material Acrolein

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit