Alfa Aesar™ Acrolein diethyl acetal, 96%

Manufacturer: Alfa Aesar™ B2482118

Quantity 50g

 View more versions of this product

Catalog No. AAB2482118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3054-95-3
    Molecular Formula CH2=CHCH(OCH2CH3)2
    Formula Weight 130.19
    MDL Number MFCD00009239
    UN Number UN2374
    Beilstein 1701567
    Synonym 3,3-Diethoxy-1-propene
    Chemical Name or Material Acrolein diethyl acetal

    Specifications

    Formula Weight 130.19
    Beilstein 1701567
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H319
    Highly flammable liquid and vapour.
    Causes serious eye irritation.
    Boiling Point 124° to 125°C
    Flash Point 13°C (55°F)
    Molecular Formula CH2=CHCH(OCH2CH3)2
    MDL Number MFCD00009239
    CAS 3054-95-3
    UN Number UN2374
    Synonym 3,3-Diethoxy-1-propene
    Quantity 50g
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 3,3-DIETHOXYPROPENE
    RTECS Number AS1370000
    Chemical Name or Material Acrolein diethyl acetal
    Assay 96%

    Description

    Applications
    Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.

    Solubility
    Slightly soluble in water.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

    GHS H Statement
    H225-H319
    Highly flammable liquid and vapour.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit