Learn More
Acrolein Mouse anti-Chemical, Clone: 2H2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527557
Catalog No. PIMA527557
Add to cart
Description
Description
Acrolein is primarily used as an intermediate in the synthesis of acrylic acid and as a biocide. Acrolein may be formed from the breakdown of certain pollutants in outdoor air or from the burning of organic matter including tobacco, or fuels such as gasoline or oil. Acrolein is toxic to humans following inhalation, oral or dermal exposures.
Specifications
Specifications
Acrolein
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Acrolein modified protein, Acrolein conjugated protein, 2-Propen-1-one, 2-propenal, Acraldehyde, Acrolein, Acrylic aldehyde, Protein-bound Acrolein
Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH)
Protein G
RUO
Antibody
Monoclonal
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
2H2
Unconjugated
Liquid
Mouse
IgG1
100 μg
-20°C
Primary
Chemical