Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrolein, Mouse anti-Human, Biotin, Clone: 2H2, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB16479

Catalog No. 89-999-462



Description

Description

Specifications

Specifications

Acrolein
2H2
ELISA (1:1000) Flow Cytometry Immunocytochemistry (1:50) Immunofluorescence (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH).
100 μg
Store at -20°C.
Primary
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Biotin
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Mouse
IgG1
RUO
Antibody
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit