Learn More
Acrolein Mouse anti-Non-species specific, FITC, Clone: 2H2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259358F
Catalog No. NBP259358F
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Acrolein Monoclonal antibody specifically detects Acrolein in Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Acrolein
2H2
ELISA, Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgG1
100 ul
Primary
Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
ELISA, Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde
Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific