Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrolein Mouse anti-Nonspecies specific, PE/Atto594, Clone: 2H2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259358PEATT594

Catalog No. N259358P594


Add to cart

Description

Description

Acrolein Monoclonal antibody specifically detects Acrolein in Nonspecies specific samples. It is validated for Flow Cytometry.
Specifications

Specifications

Acrolein
2H2
Flow Cytometry
Conjugated, Purified
Mouse
IgG1
0.1 mL
Primary
Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde
Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Nonspecies specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit