Learn More
Thermo Scientific™ AcroMetrix™ Adenovirus Controls (RUO) Available on GSA/VA Contract for Federal Government customers only.
Identify increases in random or systematic error of nucleic acid test procedures for Adenovirus DNA with these controls, which are for Research Use Only.
Manufacturer:  Thermo Scientific™ 954504
 View more versions of this product
Catalog No. 23-279-166
Add to cart
Description
Description
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of Adenovirus DNA with the Thermo Scientific™ AcroMetrix™ Adenovirus Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
AcroMetrix Adenovirus High CSF Control
High
5 x 0.5mL