Learn More
Thermo Scientific™ AcroMetrix? Citrate Plasma Dilution Matrices (RUO) Available on GSA/VA Contract for Federal Government customers only.
Minimize testing variability with the Thermo Scientific? AcroMetrix? Citrate Plasma Dilution Matrices, which are for Research Use Only.
Manufacturer:  Thermo Scientific™ 941050
 View more versions of this product
Catalog No. 23-279-171
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
Minimize testing variability with the Thermo Scientific? AcroMetrix? Citrate Plasma Dilution Matrices (RUO). These matrices provide manufacturing firms, clinical laboratories and blood banks with well-characterized, pre-screened negative diluent for use in making these preliminary dilutions. The matrices are for Research Use Only and are not for use in diagnostic procedures.
Description & Specifications
Specifications
AcroMetrix Citrate Plasma Dilution Matrix (50 mL)
50mL