Learn More
Thermo Scientific™ AcroMetrix™ CMV Controls (RUO)
Identify increases in random or systematic error of nucleic acid test procedures for human cytomegalovirus (CMV) DNA with these controls, which are for Research Use Only.
Supplier: Thermo Scientific™ 951700
Description
Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of human cytomegalovirus (CMV) DNA with the Thermo Scientific™ AcroMetrix™ CMV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
Specifications
AcroMetrix CMV Negative Control | |
Negative | |
5 x 0.8 mL |
CMV Control | |
5 x 0.8mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.