Learn More
Thermo Scientific™ AcroMetrix? EBV CSF Controls (RUO) Available on GSA/VA Contract for Federal Government customers only.
Assess the performance of molecular test procedures for EBV with the Thermo Scientific? AcroMetrix? EBV CSF Controls, which are for Research Use Only.
Manufacturer:  Thermo Scientific™ 951307A
 View more versions of this product
Catalog No. 23-279-149
Due to product restrictions, please to purchase or view availability for this product.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
Assess the analytical performance of molecular test procedures, including the DNA extraction step, for the quantitative and qualitative determination of Epstein-Barr Virus (EBV) DNA with the Thermo Scientific? AcroMetrix? EBV CSF Controls (RUO). These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for Research Use Only and are not for use in diagnostic procedures.
Description & Specifications
Specifications
AcroMetrix EBV High CSF Control
High
5 x 0.5mL