Thermo Scientific™ AcroMetrix™ EBV CSF Controls (RUO) Available on GSA/VA Contract for Federal Government customers only.
Assess the performance of molecular test procedures for EBV with the Thermo Scientific™ AcroMetrix™ EBV CSF Controls, which are for Research Use Only.
Manufacturer: Thermo Scientific™ 951304
 View more versions of this product
Catalog No. 23-279-147
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Description AcroMetrix EBV Low CSF Control
    Packaging 5 x 0.5mL
    Level Low,Low
    Assess the analytical performance of molecular test procedures, including the DNA extraction step, for the quantitative and qualitative determination of Epstein-Barr Virus (EBV) DNA with the Thermo Scientific™ AcroMetrix™ EBV CSF Controls (RUO). These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for Research Use Only and are not for use in diagnostic procedures.
  • Description & Specifications
Related Products
Product Content Feedback