Learn More
Thermo Scientific™ AcroMetrix™ EBV Plasma Controls
Assess the performance of molecular diagnostic test procedures for EBV with the Thermo Scientific™ AcroMetrix™ EBV Plasma Controls, which are for In Vitro Diagnostic Use.
$937.00 - $947.00
Specifications
Detectable Analytes | EBV |
---|---|
Type | EBV Control |
Packaging | 5 x 0.5mL |
Quantity | 5 x 0.5 mL |
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
23-279-148
|
Thermo Scientific™
961230 |
Low |
Pack of 5 for $947.00
|
|
|||||
23-279-150
|
Thermo Scientific™
961231 |
High |
Pack of 5 for $937.00
|
|
|||||
Description
Assess the analytical performance of molecular diagnostic test procedures, including the DNA extraction step, for the quantitative and qualitative determination of Epstein-Barr Virus (EBV) DNA with the Thermo Scientific™ AcroMetrix™ EBV Plasma Controls. These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for In Vitro Diagnostic Use.
Specifications
EBV | |
EBV Control | |
5 x 0.5mL | |
5 x 0.5 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.