Learn More
Thermo Scientific™ AcroMetrix™ EV Controls (RUO) Available on GSA/VA Contract for Federal Government customers only.
Assess the performance of molecular test procedures for EV with the Thermo Scientific™ AcroMetrix™ EV Controls, which are for Research Use Only.
$479.85 - $539.70
Specifications
Packaging 5 x 0.3mL
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
23-279-168 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
950500
AcroMetrix EV Low Control
Pack of 5 for $539.70
Add to cart
23-279-167 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
950499
AcroMetrix EV Negative Control
Pack of 5 for $479.85
Add to cart
Description
Description
Assess molecular test procedures, including the DNA extraction step, for the qualitative and quantitative determination of enterovirus RNA with the Thermo Scientific™ AcroMetrix™ EV Controls (RUO). The controls are for Research Use Only and are not for use in diagnostic procedures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
5 x 0.3mL