Learn More
Thermo Scientific™ AcroMetrix™ Herpes Simplex Virus Controls (RUO)
Assess the performance of molecular diagnostic test procedures for human herpes viruses such as HSV-1 and 2 with these controls, which are for Research Use Only.
$684.00 - $894.00
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
23-279-154
|
Thermo Scientific™
951300 |
AcroMetrix HHV Negative Control |
Pack of 5 for $684.00
|
|
|||||
23-279-152
|
Thermo Scientific™
951301 |
AcroMetrix HSV-1 Low Control |
Pack of 5 for $873.00
|
|
|||||
23-279-156
|
Thermo Scientific™
951302 |
AcroMetrix HSV-2 Low Control |
Pack of 5 for $876.00
|
|
|||||
23-279-157
|
Thermo Scientific™
954502 |
AcroMetrix HSV-2 High CSF Control |
Pack of 5 for $894.00
|
|
|||||
23-279-153
|
Thermo Scientific™
954501 |
AcroMetrix HSV-1 High CSF Control |
Pack of 5 for $890.00
|
|
|||||
Description
Assess the performance of nucleic acid test procedures for the quantitative and qualitative determination of Human Herpes Viruses (HHV) with the Thermo Scientific™ AcroMetrix™ Herpes Simplex Virus Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.Specifications
5 x 0.5mL | |
5 x 0.5 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.