Thermo Scientific™ AcroMetrix™ HPV Controls (RUO) Available on GSA/VA Contract for Federal Government customers only.
Assess the performance of nucleic acid test procedures for the determination of HPV DNA with these controls, which are for Research Use Only.
Manufacturer: Thermo Scientific™ 952302
 View more versions of this product
Catalog No. 23-279-134
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Description AcroMetrix HPV High Risk Positive Control (Genotype 16, for Digene HC2)
    Packaging 5 x 4mL
    Assess the performance of nucleic acid test procedures for the quantitative and qualitative determination of human papillomavirus DNA (HPV DNA) with the Thermo Scientific™ AcroMetrix™ HPV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
  • Description & Specifications
Related Products
Product Content Feedback