Learn More
Thermo Scientific™ AcroMetrix™ HPV Controls (RUO) Available on GSA/VA Contract for Federal Government customers only.
Assess the performance of nucleic acid test procedures for the determination of HPV DNA with these controls, which are for Research Use Only.
$489.83 - $886.20
Specifications
Packaging 5 x 4mL
View More Specs
Products 4
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
23-279-130 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
950075
AcroMetrix HPV-16 Genotype Control
Pack of 5 for $886.20
Add to cart
23-279-131 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
950076
AcroMetrix HPV-18 Genotype Control
Pack of 5 for $885.15
Add to cart
23-279-132 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
950077
AcroMetrix HPV-68 Genotype Control
Pack of 5 for $828.45
Add to cart
23-279-133 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
950078
AcroMetrix HPV Negative Control
Pack of 5 for $489.83
Add to cart
Description
Description
Assess the performance of nucleic acid test procedures for the qualitative determination of human papillomavirus DNA (HPV DNA) with the Thermo Scientific™ AcroMetrix™ HPV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
5 x 4mL