Learn More
Thermo Scientific™ AcroMetrix? MultiMix FFPE Control (RUO) Available on GSA/VA Contract for Federal Government customers only.
Assess performance of somatic genetic variant detection in research oncology testing with these controls, which are for Research Use Only.
Manufacturer:  Thermo Scientific™ 957186
 View more versions of this product
Catalog No. 23-279-198
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
Assess the performance of methods for the isolation, purification, and detection of somatic genetic variants in research oncology testing with the Thermo Scientific? AcroMetrix? MultiMix FFPE Controls (RUO). These controls consist of five single-use samples of a synthetic tissue that has been formalin-fixed, paraffin-embedded and sectioned to 10 microns thick. The controls are for Research Use Only and are not for use in diagnostic procedures.
Description & Specifications
Specifications
MultiMix C
5 samples
FLT3, RET, KIT, KRAS, KDR, HRAS, ERBB4, SMAD4, TP53, APC, FGFR3, PDGFRA, CSF1R, EGFR