Thermo Scientific™ AcroMetrix™ MultiMix FFPE Control (RUO) Available on GSA/VA Contract for Federal Government customers only.
Assess performance of somatic genetic variant detection in research oncology testing with these controls, which are for Research Use Only.
Manufacturer: Thermo Scientific™ 957187
 View more versions of this product
Catalog No. 23-279-199
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Target Gene FLT3, RET, KRAS, CSF1R, FGFR3, TP53, APC, KDR, STK11, PDGFRA
    Description MultiMix D
    Packaging 5 samples
    Assess the performance of methods for the isolation, purification, and detection of somatic genetic variants in research oncology testing with the Thermo Scientific™ AcroMetrix™ MultiMix FFPE Controls (RUO). These controls consist of five single-use samples of a synthetic tissue that has been formalin-fixed, paraffin-embedded and sectioned to 10 microns thick. The controls are for Research Use Only and are not for use in diagnostic procedures.
  • Description & Specifications
Related Products
Product Content Feedback