Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO) Available on GSA/VA Contract for Federal Government customers only.
Monitor assays that test for HBsAg, anti-HBc, anti-HCV, anti-HIV-1/2 and anti-HTLV-1/2 with these controls, which are for Research Use Only.
Manufacturer: Thermo Scientific™ S2409
 View more versions of this product
Catalog No. 23-279-189
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Analytes Negative Control,Negative Control
    Description AcroMetrix PeliSpy Sero Negative Control
    Packaging 10 x 5mL
    Estimate the precision and reproducibility of procedures for the detection of hepatitis B surface antigen (HBsAg), antibodies to hepatitis B core antigen (anti-HBc), antibodies to hepatitis C virus (anti-HCV), antibodies to human immunodeficiency virus type 1 group M and O (anti-HIV-1 group M and anti-HIV-1 group O), antibodies to human immunodeficiency virus type 2 (anti-HIV-2) and antibodies to human T-cell lymphotrophic virus types I and II (anti-HTLV I⁄II) with the Thermo Scientific™ AcroMetrix™ PeliSpy Serology Controls (RUO). They are for Research Use Only.
  • Description & Specifications
Related Products
Product Content Feedback