Thermo Scientific™ AcroMetrix™ Transplant Virus Panels (RUO) Available on GSA/VA Contract for Federal Government customers only.
Assess the analytical performance of molecular test procedures for transplant viruses with these panels, which are for Research Use Only.
Manufacturer: Thermo Scientific™ 942015
 View more versions of this product
Catalog No. 23-279-145
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Description AcroMetrix EBV CSF Panel
    Packaging 5 x 0.5mL
    Level 0, 1000, 10000, 100000, 1000000 IU/mL,0, 1000, 10000, 100000, 1000000 IU/mL
    Assess the analytical performance of molecular test procedures, including the nucleic acid extraction step, for the quantitative and qualitative determination of viruses related to transplant disease with the Thermo Scientific™ AcroMetrix™ Transplant Virus Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits, and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.
  • Description & Specifications
Related Products
Product Content Feedback