Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix™ VZV Controls (RUO)

Identify increases in random or systematic error of nucleic acid test procedures for human VZV DNA with these controls, which are for Research Use Only.

$868.00 - $877.00

Specifications

Type VZV Control
Packaging 5 x 0.5mL
Quantity 5 x 0.5 mL
View More Specs

Products 3
Catalog Number Mfr. No. Description Level Price Quantity & Availability  
Catalog Number Mfr. No. Description Level Price Quantity & Availability  
23-279-161
View Documents
Thermo Scientific™
954514
AcroMetrix VZV High Plasma Control High
Pack of 5 for $877.00
Add to Cart
 
23-279-160
View Documents
Thermo Scientific™
954512
AcroMetrix VZV High CSF Control High
Pack of 5 for $868.00
Add to Cart
 
23-279-159
View Documents
Thermo Scientific™
954511
AcroMetrix VZV Low CSF Control Low
Pack of 5 for $868.00
Add to Cart
 
Description

Description

Identify increases in random or systematic error of nucleic acid test procedures for the quantitative and qualitative determination of human VZV DNA with the Thermo Scientific™ AcroMetrix™ VZV Controls (RUO). Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The controls are for Research Use Only and are not for use in diagnostic procedures.

Specifications

Specifications

VZV Control
5 x 0.5mL
5 x 0.5 mL
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit