Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROS Organics™ Proteinase K

$130.40 - $197.80

Specifications

Color White
Product Type Proteinase K
Format Solid
Purity 1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC611821000
View Documents
ACROS Organics™
611821000
100mg
Each for $197.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
AC611820500
View Documents
ACROS Organics™
611820500
50mg
Each for $130.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

White
Proteinase K
Solid
1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit