Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACROS Organics Sephadex-SP C-25
Fine white powder
Manufacturer: ACROS Organics 366280100
Description
- CAS: 61840-62-8
- MFCD00132782
- Total Capacity: 2.0 to 2.6mmol Hplus/g
- Loss on Drying: 10% max.
Specifications
Glass bottle | |
10g | |
Sephadex-SP C-25 media | |
Powder |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title