Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Acrylamide, 99.5%, Molecular Biology Grade

Click to view available options
Quantity:
250g
500g
1kg
79-06-1
C3H5NO
71.079
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
acrylamide,2-propenamide,propenamide,acrylic amide,akrylamid,ethylenecarboxamide,vinyl amide,acrylic acid amide,2-propeneamide,propeneamide
6579
CHEBI:28619
prop-2-enamide
C=CC(=O)N
79-06-1
C3H5NO
71.079
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
acrylamide,2-propenamide,propenamide,acrylic amide,akrylamid,ethylenecarboxamide,vinyl amide,acrylic acid amide,2-propeneamide,propeneamide
6579
CHEBI:28619
prop-2-enamide
C=CC(=O)N
Applications
Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.

Solubility
Partly soluble in water.

Notes
Air and Light Sensitive, store in dark and away form light. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Quantity 250g
Color White
Sensitivity Air & Light Sensitive
Melting Point 82°C to 85°C
Density 1.322g/mL
Odor Odorless
Boiling Point 125°C (25mmHg)
Linear Formula CH2=CHCONH2
UN Number UN2074
Beilstein 605349
Merck Index 14,129
Formula Weight 71.08
Physical Form Powder
Percent Purity 99.5%
Grade Molecular Biology
Chemical Name or Material Acrylamide
Show More Show Less

GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement
P260-P261-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ACRYLAMIDE, SOLID

RTECSNumber : AS3325000

TSCA : TSCA