Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide: Bis-Acrylamide 19:1 (Powder/Electrophoresis), Fisher BioReagents

Click to view available options
Quantity:
100g
Packaging:
Poly Bottle
79-06-1, 110-26-9
C3H5NO
71.08
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
6579
CHEBI:28619
prop-2-enamide
NC(=O)C=C
79-06-1, 110-26-9
C3H5NO
71.08
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
6579
CHEBI:28619
prop-2-enamide
NC(=O)C=C
This dry powder mix is premixed for 5% cross-linking and is suitable for DNA sequencing and separation of low-molecular-weight proteins.
Conductivity 10μmhos max.
DNase DNase free
Electrophoresis Pass Test
UN Number 2074
Packaging Poly Bottle
Physical Form Solid
Grade Electrophoresis
Color White
Quantity 100g
Chemical Name or Material Acrylamide: Bis-Acrylamide 19:1

Recommended Storage : RT