Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acrylamide/Bis Premix, Premix Ratio: 37.5:1, MP Biomedicals
Specifications
Chemical Name or Material | Acrylamide/Bis Premix |
---|---|
Name Note | Premix Ratio: 37.5:1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN800659
|
MP Biomedicals, Inc
04800659 |
30g |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Specifications
Acrylamide/Bis Premix | |
Premix Ratio: 37.5:1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title