Acrylamide, Molecular Biology Grade, Hoefer™

Manufacturer: Hoefer, Inc GR1411X

Quantity 2 x 1kg

 View more versions of this product

Catalog No. 03-500-624

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-06-1
    Physical Form Crystalline Powder
    Assay Percent Range ≥99.9%
    Linear Formula CH2CHCONH2
    Formula Weight 71.08
    Chemical Name or Material Acrylamide
    Grade Molecular Biology Grade

    Specifications

    Physical Form Crystalline Powder
    Color White, Clear
    Quantity 2 x 1kg
    Formula Weight 71.08
    Linear Formula CH2CHCONH2
    CAS 79-06-1
    Assay Percent Range ≥99.9%
    Chemical Name or Material Acrylamide
    Grade Molecular Biology Grade

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit