Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide, PAGE, PlusOne™, GE Healthcare

Click to view available options
Quantity:
1kg
79-06-1
C3H5NO
71.079
HRPVXLWXLXDGHG-UHFFFAOYSA-N
6579
CHEBI:28619
prop-2-enamide
C=CC(=O)N
79-06-1
C3H5NO
71.079
HRPVXLWXLXDGHG-UHFFFAOYSA-N
6579
CHEBI:28619
prop-2-enamide
C=CC(=O)N
Formula Weight 71.08
Physical Form Powder
Quantity 1kg
Chemical Name or Material Acrylamide, PAGE