Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide Solution 50 ppm in 80% Methanol, Reagents

Catalog No. 01336003
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-003 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-003 Supplier Reagents Holdings Llc Supplier No. CA017300500C

Due to product restrictions, please to purchase or view availability for this product.

Acrylamide Solution 50 ppm in 80% Methanol, Reagent, use for Acrylamide analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acrylamide Solution 50 ppm in 80% Methanol
Packaging Glass Bottle
CAS 67-56-1, 7732-18-5, 7732-18-5
Specific Gravity 0.8347
Physical Form Liquid
Color Colorless
Boiling Point 68°C
Solubility Completely miscible
Type Conductivity Standard
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.