Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

79-06-1
C3H5NO
71.079
HRPVXLWXLXDGHG-UHFFFAOYSA-N
6579
CHEBI:28619

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

CAS 79-06-1
Molecular Formula C3H5NO
Packaging Poly Bottle
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES C=CC(=O)N
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.079
PubChem CID 6579
ChEBI CHEBI:28619
Formula Weight 71.08
Quantity 500g
Specific Gravity 1L = 1.12kg
Flash Point 138.00°C
Chemical Name or Material Acrylamide, Purified for Electrophoresis
Grade Ultrapure Bioreagent
Show More Show Less