Alfa Aesar™ Acrylic acid, tech. 90%, stab.

Manufacturer: Alfa Aesar™ 45779AP

Quantity 500mL

 View more versions of this product

Catalog No. AA45779AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-10-7
    Molecular Formula C3H4O2
    Linear Formula H2C=CHCO2H
    Formula Weight 72.06
    MDL Number MFCD00004367
    UN Number UN2218
    Beilstein 635743
    Merck Index 14,130
    Synonym Propenoic acid
    Chemical Name or Material Acrylic acid, Stabilized
    Grade Technical Grade

    Specifications

    Formula Weight 72.06
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 79-10-7
    Boiling Point 138° to 139°C
    Flash Point 54°C (129°F)
    Molecular Formula C3H4O2
    Linear Formula H2C=CHCO2H
    Refractive Index 1.4202
    UN Number UN2218
    Beilstein 635743
    Health Hazard 2 GHS H Statement
    H314-H226-H302-H312-H332
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Quantity 500mL
    Odor Pungent
    Physical Form Liquid
    Density 1.051g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACRYLIC ACID, STABILIZED
    TSCA TSCA
    MDL Number MFCD00004367
    Merck Index 14,130
    Synonym Propenoic acid
    Health Hazard 1 Danger
    Melting Point ≈13°C
    RTECS Number AS4375000
    Chemical Name or Material Acrylic acid, Stabilized
    Assay 90%
    Grade Technical Grade

    GHS H Statement
    H314-H226-H302-H312-H332
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit