Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Acrylic Equilateral Prisms
Experiment with diffraction, spectrum formation, and more. The Eisco™ Acrylic Equilateral Prism is ideal for studying the spectrum of light in a kinesthetically and visually effective manner.
Manufacturer: Eisco™ PH0554DC
Description
- Made with optical quality glass or clear acrylic
- Polished on all faces
- Designed for use in optical experiments
- Will create a nice rainbow
Specifications
Face Size: 25 mm | |
1 Ea. | |
100 mm |
Acrylic | |
Optics experiments and demonstrations | |
Equilateral Prism |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title