Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Acrylic Lab Workspace Partitions

Easy to clean and disinfect, these partitions help to minimize laboratory workspace proximity issues. Their clear, acrylic design allows light to pass through to the work area providing a safe, bright work environment.

$270.00 - $840.00

Specifications

Color Clear
Height (English) 24 in.
Height (Metric) 61 cm
Material Acrylic
View More Specs

Products 2
Catalog Number Style Price Quantity & Availability  
Catalog Number Style Price Quantity & Availability  
06-000-909
View Documents
Island
Each for $840.00
Add to cart
 
06-000-908
View Documents
Perimeter
Each for $270.00
Add to cart
 
Description

Description

  • Constructed from 1/4 in. acrylic
  • Include lower cord/hose passage points
  • Perimeter style partitions have triangular stabilizing feet
  • Island style partitions include stabilization feet and rear acrylic cleats for secure assembly
  • All units require some assembly
Specifications

Specifications

Clear
24 in.
61 cm
Acrylic
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit