Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSBG1 Mouse anti-Human, Clone: OTI2B5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525103

Catalog No. PIMA525103


Add to cart

Description

Description

The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.
Specifications

Specifications

ACSBG1
Monoclonal
0.95 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96GR2
BGM, KIAA0631, LPD
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23205
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2B5
Unconjugated
ACSBG1
Liquid
ACSBG1
Full length human recombit protein of ACSBG1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit