Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ACSF2 Human Recombinant Protein

Highly purified. Generating reliable and reproducible results.

$436.00 - $1298.00

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-Page
Formulation Phosphate buffered saline pH 7.4 containing 10% glycerol
Gene ID (Entrez) 80221
Molecular Weight TMW: 66.1kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25216601
View Documents
Novus Biologicals™
NBP2521660.1MG
0.1mg
Each for $436.00
Add to cart
 
NBP25216605
View Documents
Novus Biologicals™
NBP2521660.5MG
0.5mg
Each for $1,298.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 42-615 of Human ACSF2 The Recombinant Human ACSF2 Protein is derived from E. coli. The Recombinant Human ACSF2 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.5mg/mL
Phosphate buffered saline pH 7.4 containing 10% glycerol
TMW: 66.1kDa
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
ACSF2
Yes
SDS-Page
80221
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
ACSMW, acyl-CoA synthetase family member 2, AVYV493, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.26, mitochondrial, PPARG binding, long chain fatty acid acyl Co-A ligase like
Unlabeled
SDS
Product Certifications

For research use only and is not approved for use in humans or in clinical diagnosis

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit