Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549471

Catalog No. PA549471


Add to cart

Description

Description

Acyl-CoA synthases catalyze the initial reaction in fatty acid metabolism, by forming a thioester with CoA. Has some preference toward medium-chain substrates. Plays a role in adipocyte differentiation.
Specifications

Specifications

ACSF2
Polyclonal
PBS with 0.09% sodium azide
Q96CM8, Q8VCW8, Q499N5
Acyl-CoA synthetase family member 2, mitochondrial, 621-, ACSF2
Rabbit
IgG
80 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
264895, 619561, 80221
Western Blot
Unconjugated
ACSF2
Liquid
ACSF2
KLH conjugated synthetic peptide between 198-232 amino acids from the Central region of human ACSF2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit