Learn More
Abnova™ ACSL5 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00051703T01
Catalog No. 89-932-410
Add to cart
Description
Description
Specifications
Specifications
293T
NM_016234.3
pCMV-ACSL5 full-length
100 μL
Human
ACSL5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 81.4kDa
Transient overexpression cell lysate was tested with anti-ACSL5 antibody (Mfr. No. H00051703-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot