Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACSL5 Monoclonal Antibody (OTI1D4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF811872


Description
ACSL5 Monoclonal antibody specifically detects ACSL5 in Human samples. It is validated for Western BlotSpecifications
ACSL5 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P13521 | |
Mouse | |
Protein A/G | |
RUO | |
7857 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Western Blot | |
OTI1D4 | |
Unconjugated | |
SCG2 | |
1700030F05Rik; ACS2; ACS5; Acsl5; acyl-CoA synthetase 5; acyl-CoA synthetase long chain family member 5; acyl-CoA synthetase long-chain family member 5; Arachidonate--CoA ligase; FACL5; FACL5 for fatty acid coenzyme A ligase 5; fatty acid coenzyme A ligase 5; fatty acid Coenzyme A ligase, long chain 5; fatty-acid-Coenzyme A ligase, long-chain 5; HGNC:16526; LACS 5; Long-chain acyl-CoA synthetase 5; long-chain fatty acid coenzyme A ligase 5; long-chain-fatty-acid--CoA ligase 5; UNQ633/PRO1250; zgc:92083 | |
Human recombinant protein fragment corresponding to amino acids 31-617 of human SCG2 (NP_003460) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction