Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACSL6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215253
Description
ACSL6 Polyclonal antibody specifically detects ACSL6 in Mouse, Rat samples. It is validated for Western Blot.Specifications
ACSL6 | |
Polyclonal | |
Unconjugated | |
0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal | |
ACS2long-chain 6, acyl-CoA synthetase long-chain family member 6, EC 6.2.1, EC 6.2.1.3, FLJ16173, LACS 6, long fatty acyl-CoA synthetase 2, Long-chain acyl-CoA synthetase 6, long-chain-fatty-acid--CoA ligase 6 | |
Rabbit | |
68 kDa | |
0.1 mL | |
Lipid and Metabolism | |
23305 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q9UKU0 | |
ACSL6 | |
Recombinant protein encompassing a sequence within the C-terminus region of human ACSL6. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction