Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACSM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $404.50
Specifications
Antigen | ACSM1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145453
|
Invitrogen™
PA5145453 |
200 μL |
Each for $404.50
|
|
|||||
FEPA5145454
|
Invitrogen™
PA5145454 |
20 μL |
Each for $191.80
|
|
|||||
Description
ACSM1 has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). This protein acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy-and 2,3-or 3,4-unsaturated acids (in vitro). This protein functions as GTP-dependent lipoate-activating enzyme that generates the substrate for lipoyltransferase.Specifications
ACSM1 | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat, Pig | |
Q08AH1, Q91VA0 | |
116285, 117147, 361638 | |
Recombinant Human ACSM1 protein, Ser370-Pro557 (Accession #Q08AH1). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
ACSM1; Acyl-CoA synthetase medium-chain family member 1; acyl-coenzyme A synthetase ACSM1, mitochondrial; Benzoate--CoA ligase; BUCS1; Butyrate CoA ligase; butyrate--CoA ligase 1; butyryl Coenzyme A synthetase 1; butyryl-coenzyme A synthetase 1; LAE; Lipoate-activating enzyme; MACS1; medium-chain acyl-CoA synthetase; middle-chain acyl-CoA synthetase 1; Xenobiotic/medium-chain fatty acid-CoA ligase HXM-B | |
ACSM1 | |
IgG | |
Protein A | |
ACSM1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title