Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACSM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559391
Description
Immunogen sequence: KANCIITNDV LAPAVDAVAS KCENLHSKLI VSENSREGWG NLKELMKHAS DSHTCVKTKH NEIMAIFF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.
Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4-unsaturated acids (in vitro).
Specifications
ACSM3 | |
Polyclonal | |
Unconjugated | |
ACSM3 | |
Acsm3; acyl-CoA synthetase medium chain family member 3; acyl-CoA synthetase medium-chain family member 3; Acyl-coenzyme A synthetase ACSM3, mitochondrial; butyrate--CoA ligase 3; butyryl coenzyme A synthetase 3; butyryl-coenzyme A synthetase 3; CH73-60L17.3; Middle-chain acyl-CoA synthetase 3; Propionate--CoA ligase; Protein SA; protein SA homolog; Sa; SA (rat hypertension-associated) homolog; SA hypertension-associated homolog; SA rat hypertension-associated homolog; SA rat hypertension-associated protein; SAH; ssubA; zgc:172040 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6296 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53FZ2 | |
ACSM3 | |
Recombinant protein corresponding to Human ACSM3. Recombinant protein control fragment (Product #RP-98670). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction