Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSM4 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549252

Catalog No. PA549252

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with rabbit based on sequence homology.

Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4- unsaturated acids (in vitro).
Specifications

Specifications

ACSM4
Polyclonal
PBS with 0.09% sodium azide
Q80W40
Acyl-coenzyme A synthetase ACSM4, mitochondrial, Acyl-CoA synthetase medium-chain family member 4, ACSM4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
233801
Western Blot
Unconjugated
Acsm4
Liquid
ACSM4
KLH conjugated synthetic peptide between 548-576 amino acids from the C-terminal region of human ACSM4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit