Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

ACSM5 Mouse anti-Human, Clone: OTI4B11, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504490

Catalog No. 50-167-5169



Description

Description

Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4- unsaturated acids (in vitro).
Specifications

Specifications

ACSM5
Monoclonal
1.05 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6NUN0
acyl-CoA synthetase medium chain family member 5
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54988
Flow Cytometry, Western Blot
OTI4B11
Unconjugated
ACSM5
Liquid
ACSM5
Full length human recombit protein of human ACSM5 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit